Anti-STARD13 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-665
- 77845-666
- Antigen symbol:
- STARD13
- STARD13
- Antigen synonyms:
- 46H23.2|ARHGAP37|Deleted in liver cancer 2 protein|Deleted in liver cancer protein 2|DLC-2|DLC2|GT650|Rho GTPase activating protein on chromosome 13q12|Rho GTPase-activating protein|STA13|STA13_HUMAN|StAR related lipid transfer (START) domain containing 13|StAR-related lipid transfer protein 13|StARD13|START domain-containing protein 13
- 46H23.2|ARHGAP37|Deleted in liver cancer 2 protein|Deleted in liver cancer protein 2|DLC-2|DLC2|GT650|Rho GTPase activating protein on chromosome 13q12|Rho GTPase-activating protein|STA13|STA13_HUMAN|StAR related lipid transfer (START) domain containing 13|StAR-related lipid transfer protein 13|StARD13|START domain-containing protein 13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3M8
- Q9Y3M8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human StAR-related lipid transfer protein 13 protein (516-679AA)
- Recombinant Human StAR-related lipid transfer protein 13 protein (516-679AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together