Anti-CAPN2 Rabbit Recombinant Polyclonal Antibody
77852-341
New Product
- Antigen symbol:ENC1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O14682
- Antigen synonyms:CCL 28|CCL28|Ectoderm neural cortex protein 1|Ectoderm-neural cortex protein 1|Ectodermal neural cortex (with BTB like domain)|ENC 1|ENC-1|ENC1|ENC1_HUMAN|FLJ39259|Kelch like 35|Kelch like 37|Kelch-like protein 37|KLHL 35|KLHL35|KLHL37|NRPB|Nuclear matrix protein NRP/B|Nuclear restricted protein BTB domain like (brain)|OTTHUMP00000128235|OTTHUMP00000222054|p53 induced gene 10 protein|p53-induced gene 10 protein|PIG 10|PIG10|TP53I10|Tumor protein p53 inducible protein 10
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ectoderm-neural cortex protein 1 protein (1-230AA)
- Purification:>95%, Protein G purified
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human