Anti-GIT2 Rabbit Polyclonal Antibody
Catalog # 77848-413
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:GIT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q14161
- Antigen synonyms:ARF GAP GIT2|ARF GTPase activating protein GIT2|ARF GTPase-activating protein GIT2|CAT 2|CAT-2|CAT2|Cool associated tyrosine phosphorylated protein 2|Cool interacting tyrosine phosphorylated protein 2|Cool-interacting tyrosine-phosphorylated protein 2|DKFZp686G01261|G protein coupled receptor kinase interacting ArfGAP 2|G protein coupled receptor kinase interactor 2|G protein-coupled receptor kinase-interactor 2|GIT 2|GIT2|GIT2_HUMAN|GRK interacting protein 2|GRK-interacting protein 2|KIAA0148|MGC760
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human GIT2.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:3000
Type:
Antigen: GIT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat