Anti-RPA2 (phospho-Thr21) HEK293F cells Recombinant Monoclonal Antibody [clone: 3B2]
Catalog # 77850-348
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:RPA2
- Clonality:Recombinant Monoclonal
- Clone:3B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:HEK293F cells
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P15927
- Antigen synonyms:Replication protein A 32 kDa subunit|Replication factor A protein 2|RF-A protein 2|Replication protein A 34 kDa subunit|RP-A p34|RPA2|REPA2|RPA32|RPA34
- Modification:phospho-Thr21
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:A synthesized peptide derived from Human Phospho-RPA2 (T21)
- Purification:Affinity purified
Specifications
About this item
Recommended dilution: IF: 1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human