Anti-CCT2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-588
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CCT2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P78371
- Antigen synonyms:99D8.1|CCT 2|CCT beta|CCT-beta|CCTB|Chaperonin containing t complex polypeptide 1 beta subunit|Chaperonin containing t complex polypeptide 1 subunit 2|Chaperonin containing TCP1 subunit 2|Chaperonin containing TCP1 subunit 2 (beta)|CTP:phosphocholine cytidylyltransferase 2|Epididymis secretory sperm binding protein Li 100n|HEL S 100n|MGC142074|MGC142076|MGC94480|PRO1633|T complex protein 1 beta subunit|T complex protein 1 subunit beta|T-complex protein 1 subunit beta|TCP 1 beta|TCP-1-beta|TCPB_HUMAN
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-complex protein 1 subunit beta protein (206-535AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: CCT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human