Anti-DHRS3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-165
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:DHRS3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O75911
- Antigen synonyms:DD83.1|Dehydrogenase/reductase (SDR family) member 3|DHRS3|DHRS3_HUMAN|EC 1.1.1.300|MGC125166|RDH17|Retinal short chain dehydrogenase/reductase 1|Retinal short-chain dehydrogenase/reductase 1|retSDR1|Rsdr1|SDR family|member 3|SDR1|SDR16C1|Short-chain dehydrogenase/reductase 3|Short-chain dehydrogenase/reductase family
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Short-chain dehydrogenase/reductase 3 protein (30-169AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: DHRS3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse