Anti-RUVBL1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:500
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-325
- 77845-326
- Antigen symbol:
- RUVBL1
- RUVBL1
- Antigen synonyms:
- 49 kDa TATA box binding protein interacting protein|49 kDa TATA box-binding protein-interacting protein|49 kDa TBP interacting protein|49 kDa TBP-interacting protein|54 kDa erythrocyte cytosolic protein|ECP-54|ECP54|ERYTHROCYTE CYTOSOLIC PROTEIN|54-KD|INO80 complex subunit H|NMP 238|Nuclear matrix protein 238|Pontin 52|PONTIN|RuvB like 1|RuvB like AAA ATPase 1|RuvB-like 1|RUVB1_HUMAN|RUVBL1|RVB1|TAP54 alpha |TAP54-alpha|TIP49|TIP49a|TIP60 associated protein 54 alpha|TIP60-associated protein 54-alpha
- 49 kDa TATA box binding protein interacting protein|49 kDa TATA box-binding protein-interacting protein|49 kDa TBP interacting protein|49 kDa TBP-interacting protein|54 kDa erythrocyte cytosolic protein|ECP-54|ECP54|ERYTHROCYTE CYTOSOLIC PROTEIN|54-KD|INO80 complex subunit H|NMP 238|Nuclear matrix protein 238|Pontin 52|PONTIN|RuvB like 1|RuvB like AAA ATPase 1|RuvB-like 1|RUVB1_HUMAN|RUVBL1|RVB1|TAP54 alpha |TAP54-alpha|TIP49|TIP49a|TIP60 associated protein 54 alpha|TIP60-associated protein 54-alpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y265
- Q9Y265
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human RuvB-like 1 protein (101-400AA)
- Recombinant Human RuvB-like 1 protein (101-400AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together