Order Entry
Canada
ContactUsLinkComponent
Anti-MRPS2 Rabbit Recombinant Polyclonal Antibody
Anti-MRPS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MRPS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77846-139
  • 77846-138
  • Antigen symbol:
  • MRPS2
  • MRPS2
  • Antigen synonyms:
  • 28S ribosomal protein S2|28S ribosomal protein S2 mitochondrial|CGI 91|CGI91|Mitochondrial 28S ribosomal protein S2|mitochondrial|Mitochondrial ribosomal protein S2|MRP S2|MRP-S2|OTTHUMP00000022545|OTTHUMP00000022546|RT02_HUMAN|S2mt
  • 28S ribosomal protein S2|28S ribosomal protein S2 mitochondrial|CGI 91|CGI91|Mitochondrial 28S ribosomal protein S2|mitochondrial|Mitochondrial ribosomal protein S2|MRP S2|MRP-S2|OTTHUMP00000022545|OTTHUMP00000022546|RT02_HUMAN|S2mt
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y399
  • Q9Y399
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant human 28S ribosomal protein S2, mitochondrial protein (1-296AA)
  • Recombinant human 28S ribosomal protein S2, mitochondrial protein (1-296AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together