Human Urokinase (from Urine)
Catalog # CA80602-748
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:9039-53-6
- Source:Urine
- Species:Human
- Size:10 kU
- Storage conditions:Safe to freeze
- Enzyme name:Urokinase
- Enzyme synonyms:uPA