Human Recombinant Albumin (from HEK293 cells)
Supplier: Antibodies.com
Certificates
About this item
Partial length Human Albumin, expressed in HEK293 cells, suitable for ELISA, SDS-PAGE.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P02768
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:ALB
- Protein/peptide name:Albumin
- Purity:>95% by SDS-PAGE and coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 67.3 kDa after removal of the signal peptide. The apparent molecular mass of ALB-His is approximately 55 - 70 kDa due to glycosylation.
- Sequence:ALB (Asp25-Leu609) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilized from sterile phosphate buffered saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:ELISA, SDS-PAGE
Specifications
Frequently Bought Together