Order Entry
Canada
ContactUsLinkComponent
Human Recombinant MMP-13
Human Recombinant MMP-13
Catalog # CA60801-180
Supplier:  MilliporeSigma
CAS Number:  
Human Recombinant MMP-13
Catalog # CA60801-180
Supplier:  MilliporeSigma
Supplier Number:  444287-5UG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Enzyme type:
    Recombinant
  • Species:
    Human
  • Size:
    5 μg
  • Storage conditions:
    Safe to freeze, avoid repeated freeze/thaw cycles
  • Enzyme name:
    MMP-13

Specifications

About this item