Anti-ANGPTL7 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-133
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ANGPTL7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43827
- Antigen synonyms:Angiopoietin like 7|Angiopoietin like factor (CDT6)|Angiopoietin like factor|Angiopoietin related protein 7|Angiopoietin-like factor|Angiopoietin-like protein 7|Angiopoietin-related protein 7|ANGL7_HUMAN|AngX|CDT6|Cornea derived transcript 6 protein|Cornea-derived transcript 6 protein|dJ647M16.1|OTTHUMP00000001986|RP4-647M16.2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Angiopoietin-related protein 7 protein (27-296AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ANGPTL7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human