Anti-EFNB2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-549
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:EFNB2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P52799
- Antigen synonyms:EFN B2|EFNB 2|Efnb2|EFNB2_HUMAN|Eph related receptor tyrosine kinase ligand 5|EPH-related receptor tyrosine kinase ligand 5|ephrin B2|Ephrin-B2|EphrinB2|EPLG 5|EPLG5|Htk L|HTK ligand|HTK-L|HTKL|LERK 5|LERK-5|LERK5|Ligand of eph related kinase 5|MGC126226|MGC126227|MGC126228|OTTMUSP00000024973
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ephrin-B2 protein (28-229AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EFNB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human