Recombinant Human Interferon beta (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Vesphene® III st Disinfectant Cleaners
Supplier: Steris
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
Supplier: Dukal
VWR® ClikSeal Specimen Containers
Supplier: VWR International
Fendall® Saline Eyewash Bottles, Honeywell Safety
Supplier: Honeywell Safety Products
Corning® Donor Horse Serum, US Sourced, Corning
Supplier: Corning
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Forskolin ≥99% (by TLC)
Supplier: Enzo Life Sciences
Human Recombinant GDNF (from <i>E. coli</i>)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies