Anti-PADI2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: PADI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-367
- 77845-368
- Antigen symbol:
- PADI2
- PADI2
- Antigen synonyms:
- KIAA0994|OTTHUMP00000044625|PAD 2|PAD H19|PAD-H19|PAD2|PADI 2|Padi2|PADI2 protein|PADI2_HUMAN|PDI 2|PDI2|Peptidlyarginine deiminase type II|Peptidyl arginine deiminase II|Peptidyl arginine deiminase type II|Peptidylarginine deiminase II|Protein arginine deiminase|Protein arginine deiminase type 2|Protein arginine deiminase type II|Protein-arginine deiminase type II|Protein-arginine deiminase type-2
- KIAA0994|OTTHUMP00000044625|PAD 2|PAD H19|PAD-H19|PAD2|PADI 2|Padi2|PADI2 protein|PADI2_HUMAN|PDI 2|PDI2|Peptidlyarginine deiminase type II|Peptidyl arginine deiminase II|Peptidyl arginine deiminase type II|Peptidylarginine deiminase II|Protein arginine deiminase|Protein arginine deiminase type 2|Protein arginine deiminase type II|Protein-arginine deiminase type II|Protein-arginine deiminase type-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2J8
- Q9Y2J8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein-arginine deiminase type-2 protein (1-665AA)
- Recombinant Human Protein-arginine deiminase type-2 protein (1-665AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together