Anti-RAB2A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: RAB2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-135
- 77849-134
- Antigen symbol:
- RAB2A
- RAB2A
- Antigen synonyms:
- 9330148M11Rik|C80220|GTP binding protein|LHX|OTTMUSP00000004796|RAB2|RAB2 member RAS oncogene family|RAB2A|member RAS oncogene family|RAB2A_HUMAN|Ras related protein Rab 2A|RAS-associated protein RAB2|Ras-related protein Rab-2A|RP23-183L6.1|Small GTP binding protein RAB2A
- 9330148M11Rik|C80220|GTP binding protein|LHX|OTTMUSP00000004796|RAB2|RAB2 member RAS oncogene family|RAB2A|member RAS oncogene family|RAB2A_HUMAN|Ras related protein Rab 2A|RAS-associated protein RAB2|Ras-related protein Rab-2A|RP23-183L6.1|Small GTP binding protein RAB2A
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61019
- P61019
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together