Human Recombinant PP2B (from E. coli)
Catalog # CA80059-380
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:5000 U
- Storage conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Protein synonyms:CaN|Protein Phosphatase 2B|Calcineurin
- Protein/peptide name:PP2B