Recombinant Human Superoxide Dismutase (from E. coli)
Catalog # CA82603-058
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 μg
- Storage conditions:Safe to freeze
- Enzyme name:Superoxide Dismutase
- Enzyme synonyms:Cu/Zn SOD
- Purity:≥90% (SDS-PAGE)