Anti-CASP9 Rabbit Recombinant Polyclonal Antibody
77852-405
New Product
- Antigen symbol:CASP9
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P55211
- Antigen synonyms:APAF-3|Apoptotic protease Mch-6|Apoptotic protease-activating factor 3|CASP-9|CASP9_HUMAN|Caspase 9|Caspase-9 subunit p10|ICE-LAP6|ICE-like apoptotic protease 6
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Caspase-9 protein (113-324AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: CASP9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse