Anti-CYP4A11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: CYP4A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77847-251
- 77847-252
- Antigen symbol:
- CYP4A11
- CYP4A11
- Antigen synonyms:
- 20-HETE synthase|20-hydroxyeicosatetraenoic acid synthase|CP4AB_HUMAN|CYP4A|Cyp4a1|Cyp4a10|Cyp4a14|Cyp4a3|CYP4A7|CYP4AII|CYPIVA11|Cytochrome P-450HK-omega|Cytochrome P450 4A1|Cytochrome P450 4A10|Cytochrome P450 4A11|Cytochrome P450 4A14|Cytochrome P450 4A2|Cytochrome P450 4A3|Cytochrome P450 4A7|Cytochrome P450HL-omega|Fatty acid omega-hydroxylase|Lauric acid omega-hydroxylase
- 20-HETE synthase|20-hydroxyeicosatetraenoic acid synthase|CP4AB_HUMAN|CYP4A|Cyp4a1|Cyp4a10|Cyp4a14|Cyp4a3|CYP4A7|CYP4AII|CYPIVA11|Cytochrome P-450HK-omega|Cytochrome P450 4A1|Cytochrome P450 4A10|Cytochrome P450 4A11|Cytochrome P450 4A14|Cytochrome P450 4A2|Cytochrome P450 4A3|Cytochrome P450 4A7|Cytochrome P450HL-omega|Fatty acid omega-hydroxylase|Lauric acid omega-hydroxylase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CYP4A11
- Fusion protein of Human CYP4A11
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together