Human Recombinant GRO gamma (from E. coli)
Catalog # 77182-950
Supplier: Antibodies.com
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:Lyophilized: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P19876
- Reconstitution instructions:Reconstitute with distilled sterile water
- Carrier-free:N
- Protein synonyms:C-X-C motif chemokine 3|C-X-C motif chemokine ligand 3|Chemokine (C X C motif) ligand 3|Chemokine (CXC motif) ligand 3|Cinc 2|CINC 2b|Cinc2|CINC2b|CXCL 3|Cxcl3
- Protein/peptide name:GRO gamma
- Purity:>95% by SDS-PAGE under reducing conditions
- Molecular weight:10.1 KDa
- Concentration:Reconstitution dependent
- Formulation:Lyophilized from a 0.2 µm filtered solution of phosphate buffered saline, pH 7.4.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
About this item
Partial length Human GRO gamma, expressed in Escherichia coli.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.