Anti-MARCH4 Rabbit Polyclonal Antibody
Catalog # 77849-277
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:MARCH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:liquid
- Gene ID:Q9P2E8
- Antigen synonyms:MARCHF4|KIAA1399|RNF174E3 ubiquitin-protein ligase MARCHF4|EC 2.3.2.27|Membrane-associated RING finger protein 4|Membrane-associated RING-CH protein IV|MARCH-IV|RING finger protein 174|RING-type E3 ubiquitin transferase MARCHF4
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human MARCH4.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:100
Type:
Antigen: MARCH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse