Anti-ADAMTS13 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77850-747
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ADAMTS13
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:Q76LX8
- Antigen synonyms:A disintegrin and metalloproteinase with thrombospondin motifs 13|A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 13|A disintegrin like and metalloprotease with thrombospondin type 1 motif 13|ADAM metallopeptidase with thrombospondin type 1 motif 13|ADAM TS|ADAM-TS 13|ADAM-TS13|ADAMTS 13|ADAMTS-13|ADAMTS13|ADAMTS13 protein|ATS13_HUMAN|C9orf8|TTP|Von Willebrand factor cleaving protease|von Willebrand factor-cleaving protease|vWF cleaving protease|vWF CP|vWF-cleaving protease|vWF-CP|vWFCP
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 13 protein (738-905AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ADAMTS13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human