Anti-DNALI1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77854-272
New Product
- Antigen symbol:DNALI1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O14645
- Antigen synonyms:axonemal|Axonemal dynein light chain (hp28)|Axonemal dynein light chain|Axonemal dynein light intermediate polypeptide 1|dJ423B22.5|DNALI 1|DNALI1|Dynein axonemal light intermediate chain 1|Dynein axonemal light intermediate polypeptide 1|hp28|IDLC_HUMAN|Inner dynein arm homolog of clamydomonas|Inner dynein arm light chain|Inner dynein arm light chain axonemal|P28
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Axonemal dynein light intermediate polypeptide 1 protein (1-258AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: DNALI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human