Anti-STARD10 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STARD10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-630
- 77845-631
- Antigen symbol:
- STARD10
- STARD10
- Antigen synonyms:
- Antigen NY-CO-28|CGI 52|MGC14401|NY CO 28|PCTL_HUMAN|PCTP-L|PCTP-like protein|PCTP2|SDCCAG28|Serologically defined colon cancer antigen 28|StAR related lipid transfer (START) domain containing 10|StAR-related lipid transfer protein 10|StARD10|START domain containing 10|START domain-containing protein 10
- Antigen NY-CO-28|CGI 52|MGC14401|NY CO 28|PCTL_HUMAN|PCTP-L|PCTP-like protein|PCTP2|SDCCAG28|Serologically defined colon cancer antigen 28|StAR related lipid transfer (START) domain containing 10|StAR-related lipid transfer protein 10|StARD10|START domain containing 10|START domain-containing protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y365
- Q9Y365
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human START domain-containing protein 10 protein (1-291AA)
- Recombinant Human START domain-containing protein 10 protein (1-291AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together