Recombinant Human PD-L1 (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
RNA Clean & Concentrator™ Kits, Zymo Research
Supplier: Zymo Research
2-Propanol ≥99.0% USP, VWR Chemicals BDH®
Supplier: BDH
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
Orion™ pH Electrode Filling Solution, Thermo Scientific
Supplier: Thermo Fisher Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Starswab® Anaerobic Transport System, Starplex®
Supplier: Starplex
VWR® Sterile 70% Isopropanol
Supplier: VWR International
tri-Sodium citrate dihydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
Thioacetamide ≥99% ACS
Supplier: Thermo Scientific Chemicals
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Enzymes and Separation Accessories, Omega Bio-Tek
Supplier: Omega Bio-Tek
Copper(II) acetate, anhydrous
Supplier: Thermo Scientific Chemicals