Anti-ADAP1 Rabbit Recombinant polyclonal Antibody
77850-752
New Product
- Antigen symbol:ADAP1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O75689
- Antigen synonyms:CENTA1Arf-GAP with dual PH domain-containing protein 1|Centaurin-alpha-1|Cnt-a1|Putative MAPK-activating protein PM25
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Arf-GAP with dual PH domain-containing protein 1 protein (1-374AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: ADAP1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human