Recombinant Human IL-13 (from HEK293 cells)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Frequently Bought Together
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Razor Blades
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Premium Dissecting Scissors with Sharp/Blunt Blades
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Simple Nutrients Standard, NSI
Supplier: SPEX CERTIPREP LLC
Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Succinimide 98+%
Supplier: Thermo Scientific Chemicals