Anti-ALDH3A1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77851-039
New Product
- Antigen symbol:ALDH3A1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P30838
- Antigen synonyms:Acetaldehyde dehydrogenase 3|AHD 4|AHD C|AHD4|AHDC|AL3A1_HUMAN|Aldehyde dehydrogenase 3|Aldehyde dehydrogenase 3 family member A1|Aldehyde dehydrogenase 3A1|Aldehyde dehydrogenase|Aldehyde dehydrogenase class 3|Aldehyde dehydrogenase dimeric NADP preferring|Aldehyde dehydrogenase family 3 member A1|Aldehyde dehydrogenase family 3 subfamily A1|Aldehyde dehydrogenase family 3, subfamily A, member 1|Aldehyde dehydrogenase isozyme 3|Aldehyde dehydrogenase type III|ALDH 3|ALDH 3A1|ALDH III|ALDH, stomach type|ALDH3 A1|ALDH3|ALDH3A 1|Aldh3a1|ALDHIII|dimeric NADP-preferring|MGC10406|Stomach aldehyde dehydrogenase|Tumor associated aldehyde dehydrogenase tumor ALDH
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aldehyde dehydrogenase, dimeric NADP-preferring protein (210-453AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: ALDH3A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human