Anti-ATP5F1D Rabbit Recombinant Polyclonal Antibody
77851-616
New Product
- Antigen symbol:ATP5F1D
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P30049
- Antigen synonyms:ATP synthase subunit delta|mitochondrial|ATP synthase|H+ transporting|mitochondrial F1 complex|delta subunit|ATP5D|ATPD_HUMAN|F ATPase delta subunit|F-ATPase delta subunit|Mitochondrial ATP synthase complex delta subunit precusor|Mitochondrial ATP synthase delta subunit
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant human ATP synthase subunit delta, mitochondrial protein (23-168AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ATP5F1D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human