Anti-ADAMTS4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77850-750
New Product
- Antigen symbol:ADAMTS4
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75173
- Antigen synonyms:A disintegrin and metalloproteinase with thrombospondin motifs 4| A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 4| A disintegrin like and metalloprotease with thrombospondin type 1 motif 4| ADAM metallopeptidase with thrombospondin type 1 motif 4| ADAM TS 4| ADAM TS4| ADAM-TS 4| ADAM-TS4| ADAMTS 2| ADAMTS 4| ADAMTS-4| ADAMTS2| ADAMTS4| ADAMTS4 protein| ADMP 1| ADMP-1| ADMP1| Aggrecanase 1| Aggrecanase-1| Aggrecanase1| ATS4_HUMAN| KIAA0688
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 4 protein (213-319AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ADAMTS4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human