Anti-BIN1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: BIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-236
- 77848-237
- Antigen symbol:
- BIN1
- BIN1
- Antigen synonyms:
- AMPH 2|AMPH2|Amphiphysin 2|Amphiphysin II|Amphiphysin like protein|amphiphysin-like|Amphiphysin-like protein|AMPHL|Bin1|BIN1_HUMAN|Box Dependant MYC Interacting Protein 1|Box-dependent myc-interacting protein 1|Bridging integrator 1|DKFZp547F068|MGC10367|MGC105358|Myc box dependent interacting protein 1|Myc box-dependent-interacting protein 1|SH3P9
- AMPH 2|AMPH2|Amphiphysin 2|Amphiphysin II|Amphiphysin like protein|amphiphysin-like|Amphiphysin-like protein|AMPHL|Bin1|BIN1_HUMAN|Box Dependant MYC Interacting Protein 1|Box-dependent myc-interacting protein 1|Bridging integrator 1|DKFZp547F068|MGC10367|MGC105358|Myc box dependent interacting protein 1|Myc box-dependent-interacting protein 1|SH3P9
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00499
- O00499
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human BIN1
- Fusion protein of Human BIN1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together