Anti-AP1S1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77851-215
New Product
- Antigen symbol:AP1S1
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P61966
- Antigen synonyms:Adapter related protein complex 1 sigma 1A subunit| Adapter-related protein complex 1 sigma-1A subunit| Adaptor protein complex AP-1 sigma-1A subunit| Adaptor protein complex AP1 sigma 1A subunit| AP-1 complex subunit sigma-1A| AP1 complex subunit sigma 1A| AP19| AP1S1| AP1S1_HUMAN| CLAPS1| Clathrin assembly protein complex 1 sigma 1A small chain| Clathrin assembly protein complex 1 sigma-1A small chain| Clathrin coat assembly protein AP19| Golgi adaptor HA1/AP1 adaptin sigma 1A subunit| Golgi adaptor HA1/AP1 adaptin sigma-1A subunit| HA1 19 kDa subunit| Sigma 1a subunit of AP-1 clathrin| Sigma 1a subunit of AP1 clathrin| Sigma adaptin 1A| Sigma-adaptin 1A| Sigma1A adaptin| Sigma1A-adaptin
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human AP-1 complex subunit sigma-1A protein (1-127AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: AP1S1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human