Anti-RUVBL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RUVBL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-265
- 77846-266
- Antigen symbol:
- RUVBL2
- RUVBL2
- Antigen synonyms:
- 48 kDa TATA box-binding protein-interacting protein|48 kDa TBP-interacting protein|48-kDa TATA box-binding protein-interacting protein|48-kDa TBP-interacting protein|51 kDa erythrocyte cytosolic protein|CGI-46|EC=3.6.1.-|ECP-51|ECP51|Erythrocyte cytosolic protein|51-KD|INO80 complex subunit J|INO80J|MGC144733|MGC144734|MGC52995|mp47|p47|p47 protein|Repressing pontin 52|Reptin 52|REPTIN|RuvB (E coli homolog)-like 2|RUVB|E. coli|homolog-like 2|RuvB-like 2 (E. coli)|RuvB-like 2|RuvB-like protein 2|RUVB2|RUVB2_HUMAN|RVB2|TAP54-beta|TATA box-binding protein-interacting protein|48-KD|TBP-interacting protein|TIH2|TIP48|TIP49b|TIP60-associated protein 54-beta|wu:fi25f01|zreptin
- 48 kDa TATA box-binding protein-interacting protein|48 kDa TBP-interacting protein|48-kDa TATA box-binding protein-interacting protein|48-kDa TBP-interacting protein|51 kDa erythrocyte cytosolic protein|CGI-46|EC=3.6.1.-|ECP-51|ECP51|Erythrocyte cytosolic protein|51-KD|INO80 complex subunit J|INO80J|MGC144733|MGC144734|MGC52995|mp47|p47|p47 protein|Repressing pontin 52|Reptin 52|REPTIN|RuvB (E coli homolog)-like 2|RUVB|E. coli|homolog-like 2|RuvB-like 2 (E. coli)|RuvB-like 2|RuvB-like protein 2|RUVB2|RUVB2_HUMAN|RVB2|TAP54-beta|TATA box-binding protein-interacting protein|48-KD|TBP-interacting protein|TIH2|TIP48|TIP49b|TIP60-associated protein 54-beta|wu:fi25f01|zreptin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y230
- Q9Y230
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human RuvB-like 2 protein (281-444AA)
- Recombinant Human RuvB-like 2 protein (281-444AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together