Anti-GJB3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: GJB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-301
- 77848-300
- Antigen symbol:
- GJB3
- GJB3
- Antigen synonyms:
- Connexin 31| Connexin-31| Connexin31| CX 31| Cx31| CXB3_HUMAN| DFNA 2| DFNA2| DFNA2B| EKV| FLJ22486| Gap junction beta 3 protein| Gap junction beta-3 protein| Gap junction protein beta 3 31kDa| Gap junction protein beta 3| GJB 3| MGC102938
- Connexin 31| Connexin-31| Connexin31| CX 31| Cx31| CXB3_HUMAN| DFNA 2| DFNA2| DFNA2B| EKV| FLJ22486| Gap junction beta 3 protein| Gap junction beta-3 protein| Gap junction protein beta 3 31kDa| Gap junction protein beta 3| GJB 3| MGC102938
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75712
- O75712
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human GJB3
- Synthetic peptide of Human GJB3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together