Anti-CD1B Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-615
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CD1B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P29016
- Antigen synonyms:A1 domain|CD1|CD1A|CD1b antigen|CD1B antigen b polypeptide|CD1b molecule|CD1B_HUMAN|Cortical thymocyte antigen CD1A|Cortical thymocyte antigen Differentiation antigen CD1 alpha 3|hTa1 thymocyte antigen|Ly38|T cell surface glycoprotein CD1a|MGC125990|MGC125991|R1|T-cell surface glycoprotein T6/Leu6|Thymocite antigen CD1B
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-cell surface glycoprotein CD1b protein (18-303AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CD1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human