Anti-CDC34 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: CDC34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-620
- 77847-621
- Antigen symbol:
- CDC34
- CDC34
- Antigen synonyms:
- Cdc 34| Cell division cycle 34| Cell division cycle 34 homolog (S. cerevisiae)| Cell division cycle 34 homolog| E2 UB2R1_HUMAN| UBC 3| UBC3| UBCH3| UBE2 R1| UBE2R1| Ubiquitin carrier protein| Ubiquitin conjugating enzyme Ubiquitin conjugating enzyme E2 32 kDa complementing| Ubiquitin protein ligase| Ubiquitin protein ligase R1| Ubiquitin-conjugating enzyme E2 R1| Ubiquitin-conjugating enzyme E2-32 kDa complementing| Ubiquitin-conjugating enzyme E2- Ubiquitin-protein ligase R1
- Cdc 34| Cell division cycle 34| Cell division cycle 34 homolog (S. cerevisiae)| Cell division cycle 34 homolog| E2 UB2R1_HUMAN| UBC 3| UBC3| UBCH3| UBE2 R1| UBE2R1| Ubiquitin carrier protein| Ubiquitin conjugating enzyme Ubiquitin conjugating enzyme E2 32 kDa complementing| Ubiquitin protein ligase| Ubiquitin protein ligase R1| Ubiquitin-conjugating enzyme E2 R1| Ubiquitin-conjugating enzyme E2-32 kDa complementing| Ubiquitin-conjugating enzyme E2- Ubiquitin-protein ligase R1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49427
- P49427
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human CDC34
- Synthetic peptide of Human CDC34
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together