Anti-CLINT1 Rabbit Recombinant Polyclonal Antibody
77853-254
New Product
- Antigen symbol:CLINT1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q14677
- Antigen synonyms:Clathrin interacting protein|Clathrin interactor 1|Clathrin-interacting protein localized in the trans-Golgi region|CLINT 1|Clint|ENTH|Enthoprotin|EPN4|EPN4_HUMAN|Epsin 4|Epsin related protein|Epsin-4|Epsin-related protein|EpsinR
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Clathrin interactor 1 protein (1-625AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:200 to 1:3000, IHC: 1:20 to 1:200
Type:
Antigen: CLINT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat