Anti-ALDH6A1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-050
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:ALDH6A1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q02252
- Antigen synonyms:1110038I05Rik| AI314632| Aldehyde dehydrogenase 6 family member A1| Aldehyde dehydrogenase family 6 member A1| Aldehyde dehydrogenase family 6, subfamily A1| ALDH6A1| Malonate semialdehyde dehydrogenase| Malonate-semialdehyde dehydrogenase [acylating]| Methylmalonate semialdehyde dehydrogenase [acylating] mitochondrial| Methylmalonate-semialdehyde dehydrogenase [acylating]| MGC37325| MGC40271| Mitochondrial acylating methylmalonate semialdehyde dehydrogenase| mitochondrial| MMSA_HUMAN| MMSADHA| MMSDH
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial protein (274-399AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ALDH6A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse