Anti-B4GALT3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-280
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:B4GALT3
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60512
- Antigen synonyms:4-galactosyltransferase 3|4-galactosyltransferase|4-GalTase 3|b4Gal T3|b4Gal-T3|B4GT3_HUMAN|Beta 1,4 galactosyltransferase 3|Beta 1,4 GalTase 3|Beta-1|Beta-N-acetylglucosaminyl-glycolipid beta-1|Beta4Gal T3|Beta4Gal-T3|Nal synthase|UDP Gal:beta GlcNAc beta-1,4 galactosyltransferase 3|UDP-Gal:beta-GlcNAc beta-1|UDP-galactose:beta-N-acetylglucosamine beta-1
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-1,4-galactosyltransferase 3 protein (34-393AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: B4GALT3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human