Anti-CLINT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-255
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CLINT1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q14677
- Antigen synonyms:Clathrin interacting protein| Clathrin interactor 1| Clathrin-interacting protein localized in the trans-Golgi region| CLINT 1| Clint| CLINT1| ENTH| Enthoprotin| EPN4| EPN4_HUMAN| Epsin 4| Epsin related protein| Epsin-4| Epsin-related protein| EpsinR
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Clathrin interactor 1 protein (1-625AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CLINT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human