Goat Native Goat IgG (from Muscle)
Supplier: Antibodies.com
Certificates
About this item
Full length Goat Goat IgG suitable for WB, DB, ELISA, SDS-PAGE.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Muscle
- Species:Goat
- Storage conditions:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Carrier-free:N
- Protein/peptide name:Goat IgG
- Purity:>95% (SDS-PAGE)
- Concentration:10 mg/ml
- Formulation:Supplied in 0.1 M phosphate buffered saline, pH 7.4, with 0.001% sodium azide
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:WB, DB, ELISA, SDS-PAGE
Specifications
Frequently Bought Together