Anti-SIGLEC15 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: SIGLEC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-345
- 77849-344
- Antigen symbol:
- SIGLEC15
- SIGLEC15
- Antigen synonyms:
- CD33 antigen-like 3| CD33 molecule-like 3| CD33L3| HsT1361| Sialic acid-binding Ig-like lectin 15| SIG15_HUMAN| Siglec-15| SIGLEC15
- CD33 antigen-like 3| CD33 molecule-like 3| CD33L3| HsT1361| Sialic acid-binding Ig-like lectin 15| SIG15_HUMAN| Siglec-15| SIGLEC15
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZMC9
- Q6ZMC9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SIGLEC15
- Synthetic peptide of Human SIGLEC15
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together