Anti-IL22RA1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: IL22RA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-862
- 77846-861
- Antigen symbol:
- IL22RA1
- IL22RA1
- Antigen synonyms:
- CRF2 9|CRF2-9|Cytokine receptor class-II member 9|cytokine receptor classII member 9|Cytokine receptor family 2 member 9|I22R1_HUMAN|IL-22 receptor subunit alpha-1|IL-22R-alpha-1|IL-22RA1|IL22 Receptor Alpha|IL22R|IL22R1|ILTIF R1 chain|Interleukin 22 Receptor alpha 1|Interleukin 22 Receptor|Interleukin-22 receptor subunit alpha-1|ZcytoR11
- CRF2 9|CRF2-9|Cytokine receptor class-II member 9|cytokine receptor classII member 9|Cytokine receptor family 2 member 9|I22R1_HUMAN|IL-22 receptor subunit alpha-1|IL-22R-alpha-1|IL-22RA1|IL22 Receptor Alpha|IL22R|IL22R1|ILTIF R1 chain|Interleukin 22 Receptor alpha 1|Interleukin 22 Receptor|Interleukin-22 receptor subunit alpha-1|ZcytoR11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N6P7
- Q8N6P7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human IL22RA1
- Fusion protein of Human IL22RA1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together