Anti-GPR52 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50 to 1:200
Type:
Antigen: GPR52
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-601
- 77845-602
- Antigen symbol:
- GPR52
- GPR52
- Antigen synonyms:
- G protein coupled receptor 52 |GPR52|GPR52_HUMAN|Probable G protein coupled receptor 52 |Probable G-protein coupled receptor 52
- G protein coupled receptor 52 |GPR52|GPR52_HUMAN|Probable G protein coupled receptor 52 |Probable G-protein coupled receptor 52
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2T5
- Q9Y2T5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human G-protein coupled receptor 52 protein (225-265AA)
- Recombinant Human G-protein coupled receptor 52 protein (225-265AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together