Anti-KLF12 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KLF12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-395
- 77846-394
- Antigen symbol:
- KLF12
- KLF12
- Antigen synonyms:
- 2700063E05Rik|AI225908|AI315654|AP-2 repressor|AP-2rep|AP-2rep transcription factor|AP2REP|B130052C06Rik|D530033K05Rik|HSPC122|KLF12 zinc finger transcriptional repressor|KLF12_HUMAN|Krueppel like factor 12|Krueppel-like factor 12|OTTHUMP00000018503|Transcriptional repressor AP 2rep|Transcriptional repressor AP-2rep
- 2700063E05Rik|AI225908|AI315654|AP-2 repressor|AP-2rep|AP-2rep transcription factor|AP2REP|B130052C06Rik|D530033K05Rik|HSPC122|KLF12 zinc finger transcriptional repressor|KLF12_HUMAN|Krueppel like factor 12|Krueppel-like factor 12|OTTHUMP00000018503|Transcriptional repressor AP 2rep|Transcriptional repressor AP-2rep
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4X4
- Q9Y4X4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Krueppel-like factor 12 protein (36-313AA)
- Recombinant Human Krueppel-like factor 12 protein (36-313AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together