Anti-GBP2 Rabbit Recombinant Polyclonal Antibody
77855-834
New Product
- Antigen symbol:GBP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P32456
- Antigen synonyms:GBP2Guanylate-binding protein 2|EC 3.6.5.-|GTP-binding protein 2|GBP-2|HuGBP-2|Guanine nucleotide-binding protein 2|Interferon-induced guanylate-binding protein 2
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Guanylate-binding protein 2 protein (100-259AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: GBP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse