Anti-LYVE1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LYVE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-614
- 77846-615
- Antigen symbol:
- LYVE1
- LYVE1
- Antigen synonyms:
- Cell surface retention sequence-binding protein 1|CRSBP 1|CRSBP-1|CRSBP1|extracellular link domain containing 1|extracellular link domain-containing 1|Extracellular link domain-containing protein 1|HAR|Hyaluronic acid receptor|Lymphatic endothelium specific hyaluronan receptor|lymphatic vessel endothelial hyaluronan receptor 1|Lymphatic vessel endothelial hyaluronic acid receptor 1|LYVE 1|LYVE-1|LYVE1_HUMAN|XLKD1
- Cell surface retention sequence-binding protein 1|CRSBP 1|CRSBP-1|CRSBP1|extracellular link domain containing 1|extracellular link domain-containing 1|Extracellular link domain-containing protein 1|HAR|Hyaluronic acid receptor|Lymphatic endothelium specific hyaluronan receptor|lymphatic vessel endothelial hyaluronan receptor 1|Lymphatic vessel endothelial hyaluronic acid receptor 1|LYVE 1|LYVE-1|LYVE1_HUMAN|XLKD1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Y7
- Q9Y5Y7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Lymphatic vessel endothelial hyaluronic acid receptor 1 protein (20-238AA)
- Recombinant human Lymphatic vessel endothelial hyaluronic acid receptor 1 protein (20-238AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together