Anti-EGR2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-567
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:EGR2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P11161
- Antigen synonyms:AT591|CMT1D|CMT4E|DKFZp686J1957|E3 SUMO-protein ligase Early growth response 2|Early growth response protein 2|EGR-2|EGR2_HUMAN|FLJ14547|KROX 20 Drosophila homolog|Krox 20 homolog Drosophila|KROX-20|Drosophila|homolog (early growth response-2)|KROX20|Krox20 protein|Zinc finger protein Krox-20
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human E3 SUMO-protein ligase EGR2 protein (42-245AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EGR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human