Anti-ORM1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: ORM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77847-002
- 77847-001
- Antigen symbol:
- ORM1
- ORM1
- Antigen synonyms:
- A1AG1_HUMAN|AGP 1|AGP A|AGP|AGP1|Alpha 1 acid glycoprotein|Alpha-1-acid glycoprotein 1|alpha-1-AGP|Epididymis secretory sperm binding protein Li 153w|glycoprotein|alpha-1-acid|of serum|HEL S 153w|OMD 1|ORM|Orosomucoid 1|Orosomucoid-1
- A1AG1_HUMAN|AGP 1|AGP A|AGP|AGP1|Alpha 1 acid glycoprotein|Alpha-1-acid glycoprotein 1|alpha-1-AGP|Epididymis secretory sperm binding protein Li 153w|glycoprotein|alpha-1-acid|of serum|HEL S 153w|OMD 1|ORM|Orosomucoid 1|Orosomucoid-1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ORM1
- Fusion protein of Human ORM1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together